Ocimene, natural isolate
Supplier: Ricca Chemical
|
Warning
|
Synonyms:
3,7-Dimethyl-1,3,6-octatrien, 3,7-Dimethyl-1,3,6-octatriene
RCCT9196-100C6
RCCT9196-20C6
RCCT9196-750C6
RCCT9196-100C6EA
357
USD
RCCT9196-100C6
RCCT9196-20C6
RCCT9196-750C6
Ocimene, natural isolate
Ocimene
Formula:
C₁₀H₁₆ MW: 136.23 g/mol |
MDL Number:
MFCD00026437 CAS Number: 13877-91-3 UN: 2319 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...