LaMotte® Digital Topsoil Tour
Supplier: LaMotte
5973-01 R-5973-01
470354-432EA 113.2 USD
470354-432 470354-438
LaMotte® Digital Topsoil Tour
Educational Materials Environmental Educational Materials Environmental Learning Activities
The LaMotte® Topsoil tour is an exciting classroom investigation into the physical and chemical properties of soil.
  • Grades 4 to 8
  • Instructor and student manual including test procedures, data sheets and puzzles
  • Students work in groups and individually
  • Encourages critical thinking and active learning
  • Flipped learning: Students can read background material before the lesson and apply knowledge to analyze their own classroom data
  • Works on computer or tablet
  • Great for inclusion classrooms
The topsoil tour is an exciting classroom investigation into the physical and chemical properties of soil. Each student on the topsoil tour completes
seven units while conducting his/her own soil tests for soil texture, pH, nitrogen, phosphorus and potassium using simple TesTabs® reagent tablets. Incorporates math and science activities.
Activities can be modified to fit specific curriculum needs or learning opportunities. Data sheets and puzzles for each unit can be printed or filled
electronically by students using a computer or tablet. Includes instructor manual, student manual with data sheets and puzzles, classroom color
chart and reagents and test tubes for 50 students.