(2-Bromo-6-fluoro-3-isopropoxyphenyl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
15152-5G
15152-25G
77360-214EA
112.27
USD
77360-214
77372-178
(2-Bromo-6-fluoro-3-isopropoxyphenyl)boronic acid ≥97%
(2-Bromo-6-fluoro-3-isopropoxyphenyl)boronic acid
Caution: for Research Use Only
Formula:
C₉H₁₁BBrFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD08276826 CAS Number: 1072951-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...