4-Fluoro-3-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

121916-5G 121916-25G
77359-882EA 106.09 USD
77359-882 77371-864
4-Fluoro-3-methylbenzamide ≥95%
4-Fluoro-3-methylbenzamide

Caution: for Research Use Only

Formula: C₈H₈FNO
MW: 153.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631489
CAS Number: 261945-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR