1-(2,4-Dimethoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

71615-500MG 71615-1G 71615-5G
77336-944EA 400.67 USD
77336-944 77357-784 77381-710
1-(2,4-Dimethoxyphenyl)-2,2-difluoroethanone ≥97%
1-(2,4-Dimethoxyphenyl)-2,2-difluoroethanone

Caution: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD26045186
CAS Number: 1337572-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR