(2,3-Difluoro-5-methoxy-4-methylphenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
71359-500MG
71359-1G
71359-25G
71359-5G
77333-592EA
304.88
USD
77333-592
77357-724
77390-554
77381-674
(2,3-Difluoro-5-methoxy-4-methylphenyl)(methyl)sulfane ≥97%
(2,3-Difluoro-5-methoxy-4-methylphenyl)(methyl)sulfane
Caution: for Research Use Only
Formula:
C₉H₁₀F₂OS Storage Temperature: Ambient |
MDL Number:
MFCD34764098 CAS Number: 2807470-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...