5'-Fluoro-4,4''-dimethoxy-1,1':3',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

69617-500MG 69617-1G 69617-5G 69617-25G
77333-552EA 95.79 USD
77333-552 77342-592 77369-564 77381-648
5'-Fluoro-4,4''-dimethoxy-1,1':3',1''-terphenyl ≥97%
5'-Fluoro-4,4''-dimethoxy-1,1':3',1''-terphenyl

Caution: for Research Use Only

Formula: C₂₀H₁₇FO₂
Storage Temperature: Ambient
CAS Number: 2807446-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR