1-(3-Nitrophenyl)pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

69613-500MG 69613-1G 69613-5G 69613-25G
77333-548EA 89.61 USD
77333-548 77342-370 77369-560 77381-644
1-(3-Nitrophenyl)pyrrolidine ≥97%
1-(3-Nitrophenyl)pyrrolidine

Caution: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02940746
CAS Number: 132993-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR