3-Methoxy-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69417-500MG 69417-1G 69417-5G
77339-746EA 624.18 USD
77339-746 77357-678 77381-628
3-Methoxy-2,5-dimethylbenzoic acid ≥97%
3-Methoxy-2,5-dimethylbenzoic acid

Caution: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD15527594
CAS Number: 92315-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR