4-Ethoxy-2-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65038-500MG 65038-1G 65038-5G
77337-732EA 438.78 USD
77337-732 77356-738 77380-686
4-Ethoxy-2-ethylbenzoic acid ≥97%
4-Ethoxy-2-ethylbenzoic acid

Caution: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD19309495
CAS Number: 1310278-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR