1-Bromo-2-fluoro-3-iodo-5-isopropylbenzene

Supplier: AOBChem USA

Warning

64742-500MG 64742-1G 64742-5G
77338-730EA 511.91 USD
77338-730 77356-646 77380-586
1-Bromo-2-fluoro-3-iodo-5-isopropylbenzene
1-Bromo-2-fluoro-3-iodo-5-isopropylbenzene

Caution: for Research Use Only

Formula: C₉H₉BrFI
Storage Temperature: Ambient
MDL Number: MFCD34761946
CAS Number: 2807456-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR