2-Fluoro-5-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64703-500MG 64703-1G 64703-5G 64703-25G
77332-300EA 95.79 USD
77332-300 77342-580 77368-364 77380-574
2-Fluoro-5-isopropylbenzaldehyde ≥97%
2-Fluoro-5-isopropylbenzaldehyde

Caution: for Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD19598745
CAS Number: 1289064-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR