4-Bromo-2-fluoro-6-iodo-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

62149-500MG 62149-1G 62149-5G 62149-25G
77331-530EA 174.07 USD
77331-530 77346-470 77367-676 77389-118
4-Bromo-2-fluoro-6-iodo-3-methylaniline ≥97%
4-Bromo-2-fluoro-6-iodo-3-methylaniline

Caution: for Research Use Only

Formula: C₇H₆BrFIN
Storage Temperature: Ambient
MDL Number: MFCD34760738
CAS Number: 2901067-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR