2,4-Dibromo-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60764-500MG 60764-1G 60764-5G
77338-426EA 487.19 USD
77338-426 77355-236 77379-316
2,4-Dibromo-5-(trifluoromethyl)benzaldehyde ≥97%
2,4-Dibromo-5-(trifluoromethyl)benzaldehyde

Caution: for Research Use Only

Formula: C₈H₃Br₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD34759929
CAS Number: 2167801-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR