2,4-Dibromo-5-(trifluoromethoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

60738-1G 60738-5G 60738-25G
77342-026EA 111.24 USD
77342-026 77367-208 77379-302
2,4-Dibromo-5-(trifluoromethoxy)aniline ≥97%
2,4-Dibromo-5-(trifluoromethoxy)aniline

Caution: for Research Use Only

Formula: C₇H₄Br₂F₃NO
Storage Temperature: Ambient
MDL Number: MFCD28785125
CAS Number: 1806329-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR