4-Chloro-1-iodo-2-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60352-500MG 60352-1G 60352-5G
77330-860EA 324.45 USD
77330-860 77355-074 77379-150
4-Chloro-1-iodo-2-isobutoxybenzene ≥97%
4-Chloro-1-iodo-2-isobutoxybenzene

Caution: for Research Use Only

Formula: C₁₀H₁₂ClIO
Storage Temperature: Ambient
MDL Number: MFCD34759702
CAS Number: 2755719-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR