5-Acetyl-2-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
60333-500MG
60333-1G
60333-5G
77338-348EA
479.98
USD
77338-348
77355-062
77379-140
5-Acetyl-2-fluorobenzaldehyde ≥97%
5-Acetyl-2-fluorobenzaldehyde
Caution: for Research Use Only
Formula:
C₉H₇FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18824470 CAS Number: 319454-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...