6-Bromo-2-fluoro-3-propoxyphenol ≥97%

Supplier: AOBChem USA

Warning

54854-1G 54854-5G 54854-25G
77341-032EA 66.95 USD
77341-032 77365-644 77377-470
6-Bromo-2-fluoro-3-propoxyphenol ≥97%
6-Bromo-2-fluoro-3-propoxyphenol

Caution: for Research Use Only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34530320
CAS Number: 1783855-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR