(2,3-difluoro-4-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

48610-500MG 48610-1G 48610-5G 48610-25G
77327-636EA 124.63 USD
77327-636 77344-072 77363-972 77375-532
(2,3-difluoro-4-iodophenyl)(methyl)sulfane ≥97%
(2,3-difluoro-4-iodophenyl)(methyl)sulfane

Caution: for Research Use Only

Formula: C₇H₅F₂IS
Storage Temperature: Ambient
MDL Number: MFCD33403898
CAS Number: 2624417-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR