2,6-Difluoro-4-isopropyloxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31749-500MG 31749-1G 31749-5G 31749-25G
77325-298EA 148.32 USD
77325-298 77345-252 77361-654 77384-784
2,6-Difluoro-4-isopropyloxybenzaldehyde ≥97%
2,6-Difluoro-4-isopropyloxybenzaldehyde

Caution: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD30344672
CAS Number: 1781138-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR