6-Bromo-2,3-dichlorophenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
31592-500MG
31592-1G
31592-5G
31592-25G
77325-280EA
233.81
USD
77325-280
77349-012
77361-638
77384-772
6-Bromo-2,3-dichlorophenylboronic acid pinacol ester ≥97%
6-Bromo-2,3-dichlorophenylboronic acid pinacol ester
Caution: for Research Use Only
Formula:
C₁₂H₁₄BBrCl₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD29915593 CAS Number: 2121512-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...