4-(t-Butoxycarbonyl)-2-methylphenylboronic acid, pinacol ester ≥95%

Supplier: AOBChem USA

Warning

28373-500MG 28373-1G 28373-5G 28373-25G
77324-940EA 183.34 USD
77324-940 77347-112 77361-264 77384-516
4-(t-Butoxycarbonyl)-2-methylphenylboronic acid, pinacol ester ≥95%
4-(t-Butoxycarbonyl)-2-methylphenylboronic acid, pinacol ester

Caution: for Research Use Only

Formula: C₁₈H₂₇BO₄
Storage Temperature: Ambient
MDL Number: MFCD22494127
CAS Number: 1073493-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR