1-(2-Fluoro-5-bromophenyl)-N-methoxymethanimine ≥97%

Supplier: AOBChem USA

Warning

26538-1G 26538-5G 26538-25G
77341-664EA 98.88 USD
77341-664 77361-126 77372-896
1-(2-Fluoro-5-bromophenyl)-N-methoxymethanimine ≥97%
1-(2-Fluoro-5-bromophenyl)-N-methoxymethanimine

Caution: for Research Use Only

Formula: C₈H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD27756666
CAS Number: 914311-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR