4-(4-Chlorophenoxymethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21779-500MG 21779-1G 21779-5G 21779-25G
77324-338EA 172.01 USD
77324-338 77346-410 77360-660 77384-078
4-(4-Chlorophenoxymethyl)phenylboronic acid ≥97%
4-(4-Chlorophenoxymethyl)phenylboronic acid

Caution: for Research Use Only

Formula: C₁₃H₁₂BClO₃
Storage Temperature: Ambient
MDL Number: MFCD16198502
CAS Number: 1256358-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR