Sucrose Preference Test Apparatus
Supplier: MAZE ENGINEERS
4431 4432
76670-046EA 3853.34 USD
76670-046 76670-048
Sucrose Preference Test Apparatus
Animal Supplies Animal Environmental Enrichment
The Sucrose Preference Test (SPT) is a behavioral test used to assess anhedonia behaviors.
  • Acrylic construction
  • Non reflective surface
The Sucrose Preference Test (SPT) is a behavioral test used to assess anhedonia, a state characterized by the inability to experience pleasure, in small animals such as rodents. The test is based on the assumption that subjects will naturally prefer sweet solutions, such as sucrose, over plain water.