Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]

Supplier: ANTIBODIES.COM LLC
A85327-100 A85327-50
77065-188EA 555.5 USD
77065-188 77069-814
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]
Antibodies
Mouse monoclonal (3F9) antibody to CRISPR-Cas9 for WB, ICC/IF and IHC with samples derived from Streptococcus pyogenes.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB (on Cas9 transfected cells): 1:1000 to 1:1:2000, WB (on pure Cas9 protein): 1:10000 to 1:20000, ICC/IF: 1:1000 to 1:200

Type: Primary
Antigen: CRISPR-Cas9
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Streptococcus pyogenes
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR