Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 1A9]

Supplier: Antibodies.com LLC
A252413-100
76961-782EA 525 USD
76961-782
Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 1A9]
Antibodies
Mouse monoclonal [1A9] antibody to DNA Ligase 1 for WB, ELISA and IP with samples derived from Human, Mouse, Rat and Bovine.

  • Validated Applications: WB, ELISA, IP
  • Recommended Dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: DNA Ligase 1
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR