Limulus Color KY Series
Supplier: Fujifilm
291-53101
291-53601
76994-356EA
695.01
USD
76994-356
76994-376
Limulus Color KY Series
Microbiology Test Systems
Endotoxin Tests
The Limulus Color KY Series are quantitative kinetic chromogenic assays (KCA) that are endotoxin-specific and unreactive to (1,3)-ß-D-glucan. This series includes both a multi-test kit and a single-test kit, which utilize a synthetic substrate that produces a yellow color to detect endotoxin with high sensitivity.
- Reagent reacts with endotoxin over a wide range of concentrations
- Reduces the chance of interfering factors due to high sensitivity
- Can be used with the Toxinometer® or microplate reader
- KCA quantitative range detection limit of 0.0002 to 5 EU/ml for single test and 0.0005 to 5 EU/ml for multi test
- The sample size is 0.1 ml when used with tube reader; 0.05 ml sample size when used with microplate reader
All reagent kits are matched with a Control Standard Endotoxin (CSE). These reagents work well with samples that are very turbid or show difficulty in quantification by the kinetic turbidimetric assay (KTA).
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)