Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/3283]

Supplier: ANTIBODIES.COM LLC
A250250-100
76966-598EA 555.5 USD
76966-598
Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/3283]
Antibodies
Mouse monoclonal [TYRP1/3283] antibody to TRP1 for ELISA and IHC-P with samples derived from Human.

  • Validated Applications: ELISA, IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TYRP1/3283
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR