Accessories for LAL, Water
Supplier: Fujifilm
LRW-12125 LRW-2030
76994-326EA 185.52 USD
76994-326 76994-364
Accessories for LAL, Water
Microbiology Test Systems Endotoxin Tests
LAL Reagent Water can be used to fulfill all aspects of endotoxin testing. From reconstitution of both the CSE and LAL for creation of standard curves to the making of product dilutions, LAL Reagent Water provides reliable and reproducible data when paired with our endotoxin-specific LAL reagents.
  • Endotoxin-free water (<0.001 EU/mL)
  • Steam-sterilized by USP standards
  • Non-LAL reactive
  • Great for reconstitution of LAL, RSE and CSE
  • Used to perform dilution series of RSE and CSE
  • Used for sample dilutions
Available in 30 ml and 125 ml sizes