3-Ethoxypropionitrile 99%

Supplier: Ambeed
Warning

A948861-100G A948861-25G A948861-500G
77282-022EA 43 USD
77282-022 77282-024 77282-026
3-Ethoxypropionitrile 99%
3-Ethoxypropionitrile

Caution: for Research Use Only

Formula: C₅H₉NO
MDL Number: MFCD00001959
CAS Number: 2141-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR