2,2'-[Oxybis(2,1-ethanediyloxy)]bisacetic acid 98%
Supplier: Ambeed
|
Danger
|
A845076-1G
A845076-250MG
A845076-5G
77281-684EA
20
USD
77281-684
77281-686
77281-688
2,2'-[Oxybis(2,1-ethanediyloxy)]bisacetic acid 98%
2,2'-[Oxybis(2,1-ethanediyloxy)]bisacetic acid
Caution: for Research Use Only
Formula:
C₈H₁₄O₇ |
MDL Number:
MFCD00044099 CAS Number: 13887-98-4 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...