(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-SIPHOS, (S)-N,N-Dimethyl-1,2,4,5,6,7-hexahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine

A840774-100MG A840774-1G A840774-250MG
77256-244EA 103 USD
77256-244 77256-246 77256-248
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%, ee 99%
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine

Caution: for Research Use Only

Formula: C₁₉H₂₀NO₂P
MDL Number: MFCD08459340
CAS Number: 443965-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR