1-Bromotridecane 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tridecyl bromide
A806905-25G
A806905-5G
77552-484EA
34
USD
77552-484
77244-250
1-Bromotridecane 98%
1-Bromotridecane
Caution: for Research Use Only
Formula:
BrH₂C(CH₂)₁₁CH₃ MW: 263.26 g/mol |
MDL Number:
MFCD00000227 CAS Number: 765-09-3 UN: 3265 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...