1-Chlorophthalazine 95%
Supplier: Ambeed
|
Warning
|
A806675-100MG
A806675-1G
A806675-250MG
A806675-5G
77244-242EA
47
USD
77244-242
77244-244
77244-246
77244-248
1-Chlorophthalazine 95%
1-Chlorophthalazine
Caution: for Research Use Only
Formula:
C₈H₅ClN₂ |
MDL Number:
MFCD00024141 CAS Number: 5784-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...