1-Isopropenyl-4-methoxybenzene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-4-(1-propen-2-yl)benzene, 4-Isopropenylanisole

A794182-1G A794182-250MG A794182-5G
77244-170EA 80.34 USD
77244-170 77244-172 77573-544
1-Isopropenyl-4-methoxybenzene 95%
1-Isopropenyl-4-methoxybenzene

Caution: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.21 g/mol
MDL Number: MFCD09414742
CAS Number: 1712-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR