HPI-4 (Ciliobrevin A) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Hedgehog Pathway Inhibitor 4, Ciliobrevin A, 2,4-Dichloro-a-(3,4-dihydro-4-oxo-2(1H)-quinazolinylidene)-β-oxo-benzenepropanenitrile, 3-(2,4-Dichlorophenyl)-3-oxo-2-(4-oxo-3,4-dihydroquinazolin-2(1H)-ylidene)propanenitrile
A766712-10MG
A766712-1MG
A766712-25MG
A766712-50MG
A766712-5MG
77267-970EA
28
USD
77267-970
77267-972
77267-974
77267-976
77267-978
HPI-4 (Ciliobrevin A) 98%
HPI-4 (Ciliobrevin A)
Caution: for Research Use Only
Formula:
C₁₇H₉Cl₂N₃O₂ |
MDL Number:
MFCD01911757 CAS Number: 302803-72-1 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...