2-Hydroxychalcone 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Hydroxyphenyl)-1-phenylprop-2-en-1-one, 2-Hydroxybenzylidenacetophenone

A756276-100G A756276-25G A756276-5G
77267-880EA 334.75 USD
77267-880 77267-882 77267-884
2-Hydroxychalcone 98%
2-Hydroxychalcone

Caution: for Research Use Only

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
MDL Number: MFCD00016449
CAS Number: 644-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR