3-Amino-3-(4-isopropylphenyl)propionic acid 97%

Supplier: Ambeed
Warning

A726621-1G A726621-250MG
77281-012EA 26.78 USD
77281-012 77281-014
3-Amino-3-(4-isopropylphenyl)propionic acid 97%
3-Amino-3-(4-isopropylphenyl)propionic acid

Caution: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.2700 g/mol
MDL Number: MFCD02663184
CAS Number: 117391-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR