2-Imidazolidinone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Imidazolidone

A713274-100G A713274-25G
77267-652EA 8 USD
77267-652 77308-182
2-Imidazolidinone 98%
2-Imidazolidinone

Caution: for Research Use Only

Formula: C₃H₆N₂O
MDL Number: MFCD00005257
CAS Number: 120-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR