Ethyl cyclohexanecarboxylate 98%

Supplier: Ambeed
Warning

A696561-10G A696561-1G A696561-5G A696561-100G A696561-25G
77267-556EA 67 USD
77267-556 77267-558 77617-882 77617-880 77747-402
Ethyl cyclohexanecarboxylate 98%
Ethyl cyclohexanecarboxylate

Caution: for Research Use Only

Formula: C₉H₁₆O₂
MW: 156.23 g/mol
MDL Number: MFCD00041734
CAS Number: 3289-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR