3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98%

Supplier: Ambeed
Warning

A682823-100G A682823-1G A682823-25G A682823-5G
77293-154EA 871 USD
77293-154 77293-156 77293-158 77293-160
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98%
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin

Caution: for Research Use Only

Formula: C₂₀H₁₈N₂O₂S
MDL Number: MFCD00041869
CAS Number: 38215-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR