3-Hydroxythiophenol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Hydroxybenzene-1-thiol, 3-Hydroxybenzenethiol, 3-Hydroxythiophenol, 1-Hydroxy-3-mercaptobenzene, 3-Mercaptophenol, m-Hydroxythiophenol, 3-Hydroxyphenyl mercaptan
A679885-1G
A679885-25G
A679885-5G
77280-702EA
20
USD
77280-702
77569-292
77569-294
3-Hydroxythiophenol 98%
3-Hydroxythiophenol
Caution: For research use only.
Formula:
C₆H₆OS MW: 126.18 Melting Pt: 16.5 Density: 1.237 |
MDL Number:
MFCD01317829 CAS Number: 40248-84-8 UN: 3334 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...