2-Diphenylphosphino-6-methylpyridine 98%

Supplier: Ambeed
Warning

A673598-1G A673598-5G A673598-25G
77267-286EA 14 USD
77267-286 77569-282 77749-388
2-Diphenylphosphino-6-methylpyridine 98%
2-Diphenylphosphino-6-methylpyridine

Caution: For research use only.

Formula: C₁₈H₁₆NP
MDL Number: MFCD04974234
CAS Number: 132682-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR