Phenylethyl-β-D-thiogalactopyranoside 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenethylthio)tetrahydro-2H-pyran-3,4,5-triol

A654523-100MG A654523-250MG A654523-5G
77280-534EA 32 USD
77280-534 77280-536 77749-343
Phenylethyl-β-D-thiogalactopyranoside 98%
Phenylethyl-β-D-thiogalactopyranoside

Caution: for Research Use Only

Formula: C₁₄H₂₀O₅S
MDL Number: MFCD00467904
CAS Number: 63407-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR