2-Methyl-2-oxazoline 95%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-4,5-dihydrooxazole

A652744-1G A652744-25G A652744-5G
77267-178EA 5 USD
77267-178 77267-180 77267-182
2-Methyl-2-oxazoline 95%
2-Methyl-2-oxazoline

Caution: for Research Use Only

Formula: C₄H₇NO
MW: 85.11 g/mol
MDL Number: MFCD00005298
CAS Number: 1120-64-5
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR