3-Chloro-2,6-difluorophenol 98%
Supplier: Ambeed
|
Warning
|
A628924-1G
A628924-250MG
A628924-5G
77280-388EA
16
USD
77280-388
77280-390
77280-392
3-Chloro-2,6-difluorophenol 98%
3-Chloro-2,6-difluorophenol
Caution: for Research Use Only
Formula:
C₆H₃ClF₂O |
MDL Number:
MFCD01631330 CAS Number: 261762-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...