Propargyl α-D-mannopyranoside 98%

Supplier: Ambeed
Warning

Synonyms: 2-Propyn-1-yl α-D-mannoside, Propargyl ALPHA-D-mannopyranoside, 2-Propynyl α-D-mannopyranoside

A567537-100MG A567537-250MG A567537-50MG
77266-412EA 248 USD
77266-412 77266-414 77266-416
Propargyl α-D-mannopyranoside 98%
Propargyl α-D-mannopyranoside

Caution: for Research Use Only

Formula: C₉H₁₄O₆
MDL Number: MFCD19980990
CAS Number: 854262-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR