4-Methylumbelliferyl-β-D-galactopyranoside 98%

Supplier: Ambeed
Warning

A556073-5G A556073-1G
77292-468EA 177 USD
77292-468 77617-364
4-Methylumbelliferyl-β-D-galactopyranoside 98%
4-Methylumbelliferyl-β-D-galactopyranoside

Caution: for Research Use Only

Formula: C₁₆H₁₈O₈
MDL Number: MFCD00036773
CAS Number: 6160-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR