4-Hydroxy-2-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A464875-100G A464875-1G A464875-25G A464875-5G
77252-968EA 435 USD
77252-968 77252-970 77252-972 77252-974
4-Hydroxy-2-methoxybenzaldehyde 97%
4-Hydroxy-2-methoxybenzaldehyde

Caution: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD00051964
CAS Number: 18278-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR